Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Dispenser Tips for Handystep™ S Repeating Pipetter

Catalog No. 13668718 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.5 mL
1 mL
1.25 mL
10 mL
12.5 mL
2.5 mL
25 mL
5 mL
50 mL
Tip Adapters for 25 and 50 mL
Sterility:
Non-sterile
Sterile
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
13-668-718 25 mL Sterile
13-668-700 0.1 mL Non-sterile
13-668-701 0.5 mL Non-sterile
13-668-702 1 mL Non-sterile
13-668-703 1.25 mL Non-sterile
13-668-704 2.5 mL Non-sterile
13-668-705 5 mL Non-sterile
13-668-706 10 mL Non-sterile
13-668-707 12.5 mL Non-sterile
13-668-708 25 mL Non-sterile
13-668-709 50 mL Non-sterile
13-668-710 0.1 mL Sterile
13-668-711 0.5 mL Sterile
13-668-712 1 mL Sterile
13-668-713 1.25 mL Sterile
13-668-714 2.5 mL Sterile
13-668-715 5 mL Sterile
13-668-716 10 mL Sterile
13-668-717 12.5 mL Sterile
13-668-719 50 mL Sterile
13-668-720 Tip Adapters for 25 and 50 mL Non-sterile
13-668-721 Tip Adapters for 25 and 50 mL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Catalog No. 13-668-718
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Manufactured from high-quality virgin plastics for use with most standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

Manufactured from high-quality virgin plastics for use with all standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

25 mL and 50 mL tips require the use of an adapter. One reusable adapter supplied in each package of 25 mL and 50 mL tips.

  • Precision seal between the piston and cylinder ensure smooth operation, increased accuracy and precision
  • Extended piston design minimizes dead volume and maximizes fluid utilization
  • Made entirely from high-quality virgin, high density polyethylene and polypropylene (piston of 0.1 mL tip is liquid crystal polymer-LCP)

Available sterile or nonsterileFits these models but not limited to:

  • BRAND™ HandyStep™, HandyStep™ S, HandyStep™ electronic, HandyStep™ touch, HandyStep™ touch S
  • All previous Fisherbrand Repeating Pipettes
  • Rainin AutoRep™ M, AutoRep™ S, AutoRep™ E
  • Eppendorf Multipette™ 4780
  • Gilson Repetman™
  • DLAB StepMate
  • Scilogex Repeater
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 25 mL
Volume (Metric) Dispensing 5000 to 25000μL
Filtered Non-filtered
Sterility Sterile
Color Translucent
Autoclavable Not Autoclavable
Precision ≤0.4 to 0.2%
Accuracy ±≤1.5 to 0.3%
Clean Claims Free from endotoxins, DNA, RNase, DNase, and ATP
Sterilization Method Beta
Format Individually Wrapped
Tip Style Positive Displacement
Quantity 25/Pk.
Product Type Syringe Tips
Disposable Single-use
For Use With (Equipment) Most repeating pipette models (see Description), Fisherbrand HandyStep S
Graduated Yes
Material HDPE, Polypropylene
Graduations Printed
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.