Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ seripettor™ pro Bottletop Dispensers
GSA_VA

Catalog No. 13688399 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 mL
1 to 10 mL
2.5 to 25 mL
Increments:
0.04 mL
0.2 mL
0.5 mL
Viscosity Rating:
1000mm2/sec.
150mm2/sec.
75 mm2/sec.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Increments Viscosity Rating
13-688-399 1 to 10 mL 0.2 mL 150mm2/sec.
13-688-400 2.5 to 25 mL 0.5 mL 75 mm2/sec.
03-840-011 0.2 to 2 mL 0.04 mL 1000mm2/sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-688-399 Supplier BRANDTECH™ BRAND™ Supplier No. 4720440
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Each seripettor pro bottletop dispenser is supplied with discharge tube, filling tube, spare dispensing catridge, mounting tool, two polypropylene adapters (GL45/33 and GL45/38), operating manual, and one-year warranty.

Designed to handle a wider range of liquids than the regular seripettor™, including certain concentrated acids, polar solvents, and UV-sensitive media.

Key Features:

  • Easily Replaceable Wiping Seal Dispensing Cartridge: Ideal for liquids prone to crystal formation
  • Quick-Change Cartridge: Simplifies maintenance and helps keep lifetime service costs low
  • Corrosion-Resistant Pump Assembly: Features an opaque upper sleeve that shields liquids from UV light during dispensing
  • Telescoping Filling Tube: Adjusts to fit various bottle sizes without the need for cutting
  • Threaded Safety Cap: Attaches and detaches quickly with a simple twist
  • Spring-Loaded Piston: Enables one-handed, semi-automatic operation

Liquids Unsuitable for the seripettorTM Pro:

  • Hydrofluoric Acid and Similar Chemicals: Including those that corrode high-density polyethylene, PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium, and borosilicate glass
  • Carbon Disulfide
  • Liquids with High Density: Exceeding 2.2 g/cm3
  • Liquids with High Viscosity: Exceeding 300 mm2/5 (for 2 mL instrument), 150 mm2/5 (for 10 mL instrument), or 75 mm2/5 (for 25 mL instrument)
  • Liquids with Vapor Pressure up to 500 mbar

Temperature Guidelines:

  • Instrument and Reagent Temperature: Must be between 59° and 104°F (15° and 407°C)
  • Agar Culture Media: Safe up to 140°F (60°C)

Order Info

Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ≤±1.2%
Angle 90°
Product Type Seripettor Pro Bottletop Dispenser
Autoclavable Autoclavable
Color Blue/Red/White
Density Up to 2.2 g/cm3
Channels 1
Material Borosilicate Glass, Ceramic, ETFE, FEP, PFA
Volume (Metric) 1 to 10 mL
Display None
For Use With (Application) Semi-automatic dispensing of many acids, alkaline, and saline solutions
Includes Discharge tube, telescoping filling tube, spare dispensing cartridge, mounting tool, GL45/33mm and GL45/38mm polypropylene adapters, operating manual
Increments 0.2 mL
Precision ≤0.2%
Temperature (English) 59°F to 104°F
Temperature (Metric) 15°C to 40°C
Vapor Pressure Up to 500mbar
Viscosity Rating 150mm2/sec.
Warranty 1 Year
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.