Learn More
Sino Biological Human KDM1 / LSD1 Protein (His & GST Tag) 50ug
Supplier:  Sino Biological 13721H20B150
Encompass_Preferred
A DNA sequence encoding the human KDM1 (O60341-1) (Ser172-Met852) was fused with the N-terminal poly histidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.