Learn More
Sigma Aldrich N,N'-Diethyl-1,3-propanediamine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- N,nα-diethyl-1,3-propanediamine is a potential ion-exchange displacer.n,nα-diethyl-1,3-propanediamine was used as displacer in parallel batch screening for ribonuclease a and α-chymotrypsinogen a adsorbed together on high-performance sepharose sp
- It was used to develop robotic high-throughput displacer screen to identify chemically selective displacers for several protein pairs in cation exchange chromatography
Specifications
Specifications
| CAS | 10061-68-4 |
| Density | 0.819 g/mL (at 25°C (literature)) |
| Molecular Formula | C7H18N2 |
| Refractive Index | n20/D 1.4374 (literature) |
| Linear Formula | C2H5NH(CH2)3 NHC2H5 |
| Quantity | 10 g |
| MDL Number | MFCD00009036 |
| Synonym | 1,3-Bis(ethylamino)propane |
| Molecular Weight (g/mol) | 130.23 |
| Percent Purity | 97% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.