Learn More
Sino Biological Human IgG3-Fc / IGHG3 Protein 100ug
Supplier:  Sino Biological 13906HNAH100
Encompass_Preferred
A DNA sequence encoding the human IgG3 Fc region (P01860) (Glu 99-Lys 377) was expressed and purified.
Catalog No. 50-161-8898
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.