Learn More
Across International 13mm ID Round Easy-Retrieve Dry Pressing Die Set
SDP
Supplier:  Across International SDS13.K
Encompass_Preferred
A set of 13mm diameter (ID) hardened steel trapezoidal split sleeve dry pressing die set for pellet presses, ready for you to make 13mm diameter cylinder-shape pellets. These die sets are professionally built for you to work with samples that tend to stick to the inner surface of the sleeve, trapezoidal split inner sleeves with pellet ejector make sample releasing a snap.
Catalog No. 50-225-1340
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.