Learn More
Market Lab Marketlab Tube Caps, 13mm, Green
Supplier:  Market Lab 1105GN
Encompass_Preferred
The Marketlab Tube Caps 13mm are green plastic screw caps designed for 13mm diameter tubes. They feature knurled sides for easy opening and closing. The caps are made from low density polyethylene and measure 17.5mm in diameter and 11mm in height. They come in a resealable bag containing 100 caps and are manufactured in the USA. The caps are suitable for sealing a variety of 13mm tubes including those used for cosmetics, arts and crafts, adhesives, and more. They provide an air-tight seal to preserve contents.
Catalog No. NC1835745
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.