Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 14-3-3 zeta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527317 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527317 Supplier Invitrogen™ Supplier No. PA527317
  • $662.00
    $541.65 / Each of 1

    Save $120.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (89%), Pig (99%), Chicken (99%), Sheep (99%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

At least seven isoforms comprise the highly conserved 14-3-3 family of homo- and heterodimeric proteins that are abundantly expressed in all eukaryotic cells. Although more than seven isoforms of 14-3-3 have been described, some redundancies have appeared upon sequencing. The 14-3-3s are thought to be key regulators of signal transduction events mediated through their binding to serine-phosphorylated proteins. By interacting with Cdc25C, 14-3-3 regulates entry into the cell cycle and through interaction with Bad, prevents apoptosis. Other proteins that have been shown to bind to 14-3-3s include members of the protein kinase C family, Cbl, IRS-1, polyoma middle-T antigen, nitrate reductase, S-raf and the IGF-1 receptor. Detection of 14-3-3 proteins in cerebrospinal fluid has been shown to be useful in the differential diagnosis of Creutzfeldt-Jakob disease and other prion diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 zeta
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Ywhaz
Gene Accession No. P63101, P63102, P63104
Gene Alias 1110013I11Rik; 14-3-3 delta; 14-3-3 protein zeta/delta; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 zeta; 1433Z; 14-3-3z; 14-3-3zeta; 14-3-3-zeta; 143Z; AI596267; AL022924; AU020854; epididymis luminal protein 4; epididymis secretory protein Li 3; epididymis secretory protein Li 93; factor activating exoenzyme S; FAS; HEL4; HEL-S-3; HEL-S-93; KCIP-1; mitochondrial import stimulation factor S1 subunit; Msfs1; phospholipase A2; protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; SEZ-2; tyrosine 3/tryptophan 5 -monooxygenase activation protein, zeta polypeptide; tyrosine 3/tryptophan 5-monooxygenase activation protein zeta polypeptide; tyrosine 3-monooxygenase tryptophan 5-monooxygenase activation protein; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, delta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide; YWHAD; YWHAZ
Gene Symbols Ywhaz
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human 14-3-3 zeta. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22631, 25578, 7534
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.