Learn More
REVIVAL ANIMAL HEALTH 14 FRENCH 16IN FEEDING TUBE
Supplier:  REVIVAL ANIMAL HEALTH 252201437
Encompass
14 French 16in feeding tube, Flared tip fits on a regular luer syringeRounded tips prevent injury to the animal
Catalog No. NC2132530
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.