Learn More
Description
- Comes with straight fiberglass handle with ribbed grip for better control.
- Made in the USA out of beryllium-free alloy.
- Built for spark-free striking in sensitive or hazardous workplace settings.
- Length 14 in.
Specifications
Specifications
| Product Type | Double-faced Hammer |
| Length (English) | 14 in. |
| Weight (English) | 2.00 lb. |
| Weight (English) Head | 1.75 lb. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
