Learn More
Cayman Chemical 1400W (hydrochloride), 214358-33-5, 10 mg
SDP
Supplier:  Cayman Chemical 8152010
Encompass_Preferred
Molecular Formula C10H15N3 • 2HCl, Purity ≥98%, Formula Weight 250.2, Synonyms: N-[[3-(aminomethyl)phenyl]methyl]-ethanimidamide, dihydrochloride; 1400W is a potent, selective inhibitor of iNOS.{4277} Using purified human iNOS, nNOS, and eNOS, 1400W exhibits Ki values of 7 nM, 2 M, and 50 M, respectively.{4277} In rat aortic rings it is at least 1,000-fold more potent against iNOS compared to eNOS.{4277} In mice bearing a murine mammary carcinoma, continuous infusion of 1400W at a rate of 10 mg/kg for 6 days decreased tumor weight approximately 50% compared to controls.{5066}.
Catalog No. NC0190933
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.