Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 1400W, iNOS Inhibitor, Selective, Solid, 10 mg

Catalog No. 502584662
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4662 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4662 Supplier Abcam Supplier No. AB12016510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective iNOS inhibitor with 5,000-fold selectivity over eNOS.

  • Activity: Selective iNOS inhibitor (5000-fold selective over eNOS)
  • Potency: Kd values of ≤7 nM, 2 μM, and 50 μM at human iNOS, nNOS, and eNOS, respectively
  • Molecular Weight: 250.17 Da
  • Purity: >97%
  • Physical Form: Solid
  • Composition: 100% N'-[[3-(aminomethyl)phenyl]methyl]ethanimidamide;dihydrochloride

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Content And Storage Store at 4°C.
Buffer 100% N'-[[3-(aminomethyl)phenyl]methyl]ethanimidamide;dihydrochloride
Target Molecule iNOS, nNOS, eNOS
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Cross Reactivity Human
Formulation 100% N'-[[3-(aminomethyl)phenyl]methyl]ethanimidamide;dihydrochloride
Quantity 10 mg
For Use With (Application) iNOS inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.