Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 1400W, Selective iNOS Inhibitor, Solid, 50 mg

Catalog No. 502584661
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4661 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4661 Supplier Abcam Supplier No. AB12016550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly selective iNOS inhibitor with over 5,000-fold selectivity over eNOS.

  • Purity: >97%
  • Molecular Weight: 250.17 Da
  • Mechanism: Selective iNOS inhibitor (5000-fold selective over eNOS)
  • Binding Affinity: Kd values are ≤7 nM, 2 μM and 50 μM at human iNOS, nNOS and eNOS, respectively
  • Chemical Name: N'-[[3-(aminomethyl)phenyl]methyl]ethanimidamide;dihydrochloride

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule iNOS
Shipping Condition Ambient
Reactivity Human
Buffer 100% N'-[[3-(aminomethyl)phenyl]methyl]ethanimidamide; dihydrochloride
Content And Storage Store at 4°C.
Form Solid
Cross Reactivity Human
Formulation 100% N'-[[3-(aminomethyl)phenyl]methyl]ethanimidamide;dihydrochloride
Quantity 50 mg
For Use With (Application) iNOS inhibition
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.