Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4,6-Trimethylpyridine

Catalog No. 142387500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-238-7500ML 500 mL
14-238-7100ML 100 mL
14-238-71L 1 L
14-238-75ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-238-7500ML Supplier Sigma Aldrich Supplier No. 142387500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,4,6-Trimethylpyridine

Specifications

CAS 108-75-8
Melting Point -43°C
Density 0.917 g/mL at 25°C
Boiling Point 171°C to 172°C
Molecular Formula C8H11N
Refractive Index n20/D 1.498
Linear Formula C8H11N
MDL Number MFCD00006338
Quantity 500 mL
Synonym 2,4,6-Collidine; sym-Collidine
Molecular Weight (g/mol) 121.18 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.