Learn More
Sino Biological Human IL1RL2 / IL-1Rrp2 Protein (His Tag) 100ug
Supplier:  Sino Biological 14398H08H100
Encompass_Preferred
A DNA sequence encoding the human IL1RL2 (NP_003845.2) (Met1-Tyr337) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.