Learn More
ATCC 143B; Osteosarcoma; Human
Supplier:  ATCC CRL8303
Encompass
143B is a mixed cell that was isolated from the bone of a 13-year-old, White female with osteosarcoma. The cell is a suitable transfection host and can be used in cancer research
Catalog No. 50-238-3147
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.