Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Hexanedione

Catalog No. 14416925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-416-925G 25 g
14-416-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-416-925G Supplier Sigma Aldrich Supplier No. 14416925G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3-hexanedione is a major component of the male-produced aggregation pheromone of anelaphus inflaticollis chemsak
  • It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (n4) ligand.2,3-hexanedione was used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems

Specifications

CAS 3848-24-6
Density 0.934 g/mL (at 25°C (literature))
Boiling Point 128°C (lit.)
Molecular Formula C6H10O2
Linear Formula CH3CH2CH2COCOCH3
Refractive Index n20/D 1.412 (literature)
MDL Number MFCD00009398
Quantity 25 g
Synonym Acetyl butyryl
Molecular Weight (g/mol) 114.14
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.