Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-alpha-Bisabolol

Catalog No. 144625ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-462-5ML 5 mL
14-462-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-462-5ML Supplier Sigma Aldrich Supplier No. 144625ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • (-)-α-bisabolol is the major constituent of essential oil from leaves of hymenocrater yazdianus.(-)-α-bisabolol was used to investigate the leishmanicidal and cytotoxic activity of essential oil of vanillosmopsis arborea.α-bisabolol is active against primary acute leukemia cells, including bcr-abl(+) acute lymphoblastic leukemias
  • It is the inhibitor of voltage-dependent ca(2+) channels in tracheal smooth muscle preparations of rat
  • It also inhibits human and rat glioma

Specifications

CAS 23089-26-1
Molecular Formula C15H26O
Refractive Index n20/D 1.496
Linear Formula C15H26O
MDL Number MFCD03412455
Quantity 5 mL
Synonym (-)-alpha-Bisabolol; (-)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol; Levomenol
Molecular Weight (g/mol) 222.37
Percent Purity ≥93% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.