Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,7-Dibromoheptane

Catalog No. 14499150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-499-150G 50 g
14-499-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-499-150G Supplier Sigma Aldrich Supplier No. 14499150G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,7-dibromoheptane was used in the synthesis of random thermotropic liquid crystalline copolyether and ternary copolyether
  • It was also used in the synthesis of thermotropic liquid crystalline dendrimer exhibiting a nematic phase

Specifications

CAS 4549-31-9
Density 1.51 g/mL (at 25°C (literature))
Boiling Point 255°C (lit.)
Molecular Formula C7H14Br2
Refractive Index n20/D 1.503 (literature)
Linear Formula Br(CH2)7 Br
MDL Number MFCD00000274
Quantity 50 g
Synonym Heptamethylene dibromide
Molecular Weight (g/mol) 257.99
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.