Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isovaleraldehyde

Catalog No. 146455100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-645-5100ML 100 mL
14-645-525ML 25 mL
14-645-5500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-645-5100ML Supplier Sigma Aldrich Supplier No. 146455100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isovaleraldehyde is an attractant and its interaction with zoospores of the fungus phytophthora palmivora has been studied by binding techniques.isovaleraldehyde was used as standard to evaluate the quality of olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis
  • It was used as internal standard in determination of wine aroma carbonyl compounds with 5-8 carbon atoms

Specifications

CAS 590-86-3
Density 0.803 g/mL (at 25°C (literature))
Boiling Point 90°C (lit.)
Molecular Formula C5H10O
Refractive Index n20/D 1.388 (literature)
Linear Formula (CH3)2 CHCH2CHO
MDL Number MFCD00007014
Quantity 100 mL
Synonym 3-Methylbutanal; 3-Methylbutyraldehyde; NSC 404119
Molecular Weight (g/mol) 86.13
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.