Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3,5-Benzenetricarbonyl trichloride

Catalog No. 14753212500 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
12 x 500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-753-212500 12 x 500 g
14-753-2100G 100 g
14-753-225KG 2.5 kg
14-753-2500G 500 g
14-753-210G 10 g
14-753-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-753-212500 Supplier Sigma Aldrich Supplier No. 14753212X500GGL
Add to Cart
Add to Cart
Request a Quote

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3,5-Benzenetricarbonyl trichloride

Specifications

CAS 4422-95-1
Melting Point 32°C to 38°C
Density 1.487 g/mL (at 25°C)
Boiling Point 180°C (6 mmHg)
Molecular Formula C9H3Cl3O3
Linear Formula C6H3(COCl)3
Quantity 12 x 500 g
MDL Number MFCD00000679
Synonym Benzene-1,3,5-tricarbonyl chloride; Trimesic acid trichloride; Trimesoyl chloride
Molecular Weight (g/mol) 265.48
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.