Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Air-Tite™ Sterile Syringes

Catalog No. 14817179 Shop All Air-Tite Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
30 mL
60 mL
Product Type:
Luer Lock
Luer Slip
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Product Type
14-817-179 1 mL Luer Slip
14-817-109 3 mL Luer Lock
14-817-180 5 mL Luer Slip
14-817-181 10 mL Luer Slip
14-817-110 5 mL Luer Lock
14-817-182 60 mL Luer Slip
14-817-111 20 mL Luer Lock
14-817-210 3 mL Luer Slip
14-817-112 30 mL Luer Lock
14-817-113 60 mL Luer Lock
14-817-115 30 mL Luer Slip
14-817-108 1 mL Luer Lock
14-817-178 10 mL Luer Lock
14-817-114 20 mL Luer Slip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 14-817-179 Supplier Air-Tite™ Supplier No. MS1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Air-Tite™ Sterile Syringes are high quality, yet economical for laboratory and veterinary use only

  • Smooth plunger
  • Bold graduations
  • Disposable
  • Sterile
  • Polypropylene
  • Economical
  • Single-use
  • Latex Free
  • ISO Compatible

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 13485
Material Polyisoprene Rubber (Tip), Polypropylene, Polypropylene (Barrel), Polypropylene (Plunger)
Color Clear
For Use With (Application) Laboratory / Veterinary
Latex Free Yes
Product Type Luer Slip
Disposable Single-use
Packaging Individually Wrapped
Volume (Metric) 1 mL
Connection Type Leur Slip
Graduated Yes
Sterility Sterile
Show More Show Less

Not for human use - laboratory and veterinary use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.