Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Homovanillyl alcohol

Catalog No. 14883010G
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-883-010G 10 g
50-185-5066 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-883-010G Supplier Sigma Aldrich Supplier No. 14883010G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Homovanillyl alcohol

Specifications

CAS 2380-78-1
Melting Point 40°C to 42°C (lit.)
Molecular Formula C9H12O3
Linear Formula HOC6H3(OCH3)CH2CH2OH
Quantity 10 g
MDL Number MFCD00002903
Synonym 4-Hydroxy-3-methoxyphenethanol; 4-Hydroxy-3-methoxyphenethyl alcohol
Molecular Weight (g/mol) 168.19
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.