Learn More
Cayman Chemical 15-deoxy-12,14-Prostaglandin J2, 87893-55-8, 1 mg
SDP
Supplier:  Cayman Chemical 18570.11
Encompass_Preferred
Molecular Formula C20H28O3, Purity ≥95% (mixture of isomers), Formula Weight 316.4, Synonyms: 11-oxo-prosta-5Z,9,12E,14E-tetraen-1-oic acid, 15-deoxy-Δ12,14-PGJ2; This formulation of 15-deoxy-.DELTA.12,14-PGJ2 contains 90-95% of the trans,trans-.DELTA.12,14 isomer, and 5-10% of other double bond isomers which have similar PPAR ligand activity (see Catalog No. 18570).
Catalog No. NC0897616
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.