Learn More
Justrite™ 15 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822323 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Light Neutral
Silver
Yellow
Door Style:
Left-hinged, Manual-close
Right-hinged, Manual-close
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Door Style
19-822-323 Silver Right-hinged, Manual-close
19-822-320 Silver Left-hinged, Manual-close
19-822-322 Yellow Left-hinged, Manual-close
19-822-324 Yellow Right-hinged, Manual-close
19-822-325 Light Neutral Left-hinged, Manual-close
19-822-326 Light Neutral Right-hinged, Manual-close
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-822-323 Supplier Justrite™ Supplier No. 882404
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 15 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Durable poly in safety Red or translucent white to easily detect liquid level
  • Current-carrying, patented carbon insert embedded in the container allows for proper grounding
  • HDPE disposal cans provide superior chemical resistance and leak protection
  • Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Door Style Right-hinged, Manual-close
Load Bearing Capacity 15 gal.
Color Silver