Learn More
Justrite™ 15 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors
Shop All Justrite Safety Group Products
Description
- Durable poly in safety Red or translucent white to easily detect liquid level
- Current-carrying, patented carbon insert embedded in the container allows for proper grounding
- HDPE disposal cans provide superior chemical resistance and leak protection
- Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Door Style | Right-hinged, Manual-close |
| Load Bearing Capacity | 15 gal. |
| Color | Yellow |