Learn More
Leica Biosystems 15 mL Bond Anti-Fluorescein Ab
Supplier:  Leica Biosystems AR0222
Encompass
ApplicationIn situ hybridization (ISH) allows the detection and visualization of specific nucleic acids in tissues sections. ISH probes used for the detection of mRNA or DNA on BOND contain a fluorescein label. The BOND AntiFluorescein Antibody allows linking of the oligonucleotide probe with the detection reagents, and consequently, visualization of a chromogenic product by light microscopy.ComponentsBOND Anti-Fluorescein Antibody is a purified IgG fraction of a mouse monoclonal antibody. It is supplied ready-to-use.
Catalog No. 50-255-2371
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.