Learn More
Cayman Chemical 15 R-ProstaglndIn F2a 10mg
SDP
Supplier:  Cayman Chemical 1674010 MG
Encompass_Preferred
An orally active small molecule AdipoR agonist that binds AdipoR1 and AdipoR2 (KDs 1.8 and 3.1 UM respectively) at 50 mg/kg activates AMPK and PPARa pathways and reduces insulin resistance and glucose intolerance in wild type mice fed a high-fat diet as well as in genetically obese db/db mice
Catalog No. 50-296-5194
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.