Learn More
Leica Biosystems Metal Base Molds 15 x 15 x 6 mm (Pack of 10)
Supplier: Leica Biosystems 3803081
Reusable metal base molds used for forming embedding wax blocks in histology workflows. Each mold measures 15 x 15 x 6 mm and is supplied as a pack of 10. The molds fit standard tissue embedding cassettes and provide consistent block formation and efficient heat transfer.
- Reusable metal construction for repeated use.
- Internal rounded corners allow easy removal of formed wax blocks.
- Dimensions: 15 x 15 x 6 mm.
- Pack quantity: 10 per pack.
- Compatible with most standard tissue embedding cassettes.
- Provides rapid heat transfer for efficient wax setting.