Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 1500 Series Class II Type B2 Bio Safety Cabinets

Catalog No. 13201871
Change view
Click to view available options
Plug Type:
North America (115 V, 15 A)
Width (English) Cabinet:
4 ft.
6 ft.
Dimensions (L x W x H) Exterior:
31.6 x 54.3 x 72.6 in.
31.6 x 78 x 72.6 in.
Lighting:
With UV light
Without UV light
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Width (English) Cabinet Dimensions (L x W x H) Exterior Lighting
13-201-871 North America (115 V, 15 A) 6 ft. 31.6 x 78 x 72.6 in. Without UV light
13-201-866 North America (115 V, 15 A) 4 ft. 31.6 x 54.3 x 72.6 in. With UV light
13-201-862 North America (115 V, 15 A) 6 ft. 31.6 x 78 x 72.6 in. With UV light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-201-871 Supplier Thermo Scientific™ Supplier No. 1512B2
Add to Cart
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Our Class II, Type B2 Biological Safety Cabinet is designed for applications that require greater dilution of volatile toxic chemicals or the aseptic processing of hazardous drugs.

Thermo Scientific™ 1500 Series Class II, Type B2 BSC is available in nominal widths of 4 ft (1.2 m) and 6 ft (1.8 m) and is fully compliant with NSF/ANSI 49 standards. It features an ergonomic design, advanced DC motor technology, and a convenient touchscreen user interface allows for easy monitoring of cabinet conditions.

  • NSF/ANSI 49 listed Class II, Type B2 BSC
  • Auto compensation for downflow, allowing for safe use of air pre-filters for additional extension of HEPA filter life
  • Advanced DC motor
  • Stainless steel interior
  • 5 in. touchscreen display for visualizing and monitoring cabinet conditions, including, airflow status, alerts and alarms, UV light timer, HEPA filter life indicator, Data logging, and Connectivity
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NSF Class II Type B2 BSC, NSF/ANSI 49, UL, CE
Chamber Material Stainless Steel (304)
Product Type Biosafety Cabinet
Model 1500 Series B2
Noise Level 66 dBA
Plug Type North America (115 V, 15 A)
Receptacles 2 Duplex, GFI
Sash Opening (Metric) 20.3 cm
Width (English) Exterior 78 in.
Width (Metric) Exterior 198.1 cm
Lighting Without UV light
Sash Opening (English) 8 in.
Dimensions (L x W x H) Exterior 31.6 x 78 x 72.6 in.
Width (English) Interior 72.5 in.
Width (English) Cabinet 6 ft.
Electrical Requirements 115 V, 50/60 Hz
Voltage 115 V
Energy Consumption (Operating Set Point) 0.37 kW
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.