Learn More
3M™ 1500-Series Safety Sunwears
Catalog No. 17623757 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Black
Black, Silver
White
Lens Tint:
Gray
Silver Mirror
Lens Coating:
Anti-fog
Scratch-resistant
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
17-623-757 Black Gray -
17-623-755 Black, Silver Silver Mirror Scratch-resistant
17-623-754 White Gray Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-623-757 Supplier 3M™ Supplier No. 7010315319
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Perfectly blending comfort, performance and flair, the 1500-Series offers a streamlined frame design with attitude and bold style. The removable foam-lined gasket helps provide cushioned protection against impact, sun, wind, and nuisance dust.
3M™ Safety Sunwear Protective Eyewear helps to protect eyes from dust, debris and the harmful rays of the sun, yet look bold and contemporary for a stylish appearance. These safety eye glasses combine the protection you need with the appearance you want.
  • On-the-job eye protection with a sleek and bold design
  • Removable foam liners offer extra comfort and protection from nuisance dust
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • Meets the requirements of CSA Z94.3 and ANSI Z87.1-2010
  • Anti-fogging properties help maintain visibility and reduce the need to remove glasses to wipe away moisture from the lenses
  • 3M Catalog# SS1514AS-B, SS1511AF-B, SS1514AS-S, SS1502AF-B
TRUSTED_SUSTAINABILITY
Specifications
Lens Tint Gray
Product Type Safety glasses
For Use With (Application) Eye protection
Frame Color Black
Lens Material Polycarbonate
Frame Type Streamline design; Full-frame style