Learn More
Nanocomposix BioReady Gold Nanoshells - Streptavidin - 150 nm, 10 OD in aqueous conjugation buffer, 5 mL
SDP
Supplier:  Nanocomposix GSIR1505M
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.150 nm streptavidin-conjugated gold nanoshells combine strong plasmonic optical properties with readily biofunctionalized surface chemistry. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC2885932