Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Distilled Water
GREENER_CHOICE

Catalog No. 15230162
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
10 x 500 mL
20 x 100 mL
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-230-162 500 mL
15-230-170 100 mL
15-230-147 1000 mL
15-230-196 20 x 100 mL
15-230-204 10 x 500 mL
15-230-001 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15-230-162 Supplier Gibco™ Supplier No. 15230162
Add to Cart
Edge
Add to Cart

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents

This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, this distilled water features:

  • Cell culture quality
  • Proven performance
  • Suitable for use in cell culture and molecular biology applications

Cell culture quality

This water is prepared by distillation and filtration (0.1 μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly.

Proven performance

The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.

Use in cell culture and molecular biology applications

This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers.

Dual-site cGMP manufacturing and quality system

This water is produced at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical product made in our Grand Island facility (15230-162). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Grade Cell Culture
Packaging Bottle
Sterility Sterile-filtered
Treatment(s) Not DEPC-Treated
Quantity 500 mL
Purification Method Membrane-Filtered
Shipping Condition Room Temperature
Is Distilled Water (Cat. No. 15230147) RNAse-free?

Distilled Water (Cat. No. 15230147) is not RNAse-free. Instead, we would recommend using UltraPure DNase/RNase-Free Distilled Water (Cat. No. 10977049) to have it DNase/RNase-free.

Can I get the conductivity measurement for Distilled Water?

We do not test the “finished” product for conductivity as part of the QC testing specifications. However, conductivity testing is done in our water system on a daily basis. This is the water system used in the manufacture of cell culture media and reagents made at the Gibco site.

Conductivity testing is performed according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.

What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?

These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.