Learn More
Branson Ultrasonics™ Ultrasonic Generator System Accessories
Catalog No. 15335120 Shop All Branson Ultrasonics Corporation Products
Change view
Click to view available options
For Use With (Equipment):
7000 Series 22 Gal. Tank
7000 Series 5.2 Gal. Tank
Material:
Wire Mesh No. 3
Wire Mesh No. 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material
15-335-120 7000 Series 5.2 Gal. Tank Wire Mesh No. 3
15-335-122 7000 Series 22 Gal. Tank Wire Mesh No. 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-335-120 Supplier Branson Ultrasonics™ Supplier No. CPN916023
Add to Cart
Add to Cart
Basket for parts to sit safely into the ultrasonic tank without touching tank bottom.
  • All 304 stainless steel construction makes baskets compatible with most cleaning processes
  • Type 304 stainless steel is acceptable for use with all alkaline solutions and many acids
  • Basket has Electro polished finish
  • Carryover and dragout of cleaning solution are minimized by open design and construction
Specifications
Type Mesh Basket
For Use With (Equipment) 7000 Series 5.2 Gal. Tank
Material Wire Mesh No. 3