Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Pre-Filled Bead Mill Tubes
GREENER_CHOICE

For use with Bead Mill 4 and Bead Mill 24 homogenizers. May also be used with most other commercial Bead Mills.

 View more versions of this product

Catalog No. 15-340-156


Only null left
Edge
Add to Cart

Includes: 50 7mL tubes pre-filled with 1.4mm ceramic beads

Description

Description

  • Used for processing brain, liver, kidney, lung, spleen
  • 7mL polypropylene tubes closed with screw cap
  • Can be autoclaved at 121°C
  • Max centrifuge speed of 5,000 x g
  • Processing range: 1mL to 5mL
  • Non-Sterile
Specifications

Specifications

Soft Tissue Homogenizing Mix Tubes
1.4 mm
15.3 mm
Ceramic
Bead Mill 24 Homogenizer
7 mL
1 to 5 mL
57 mm (without cap); 60 mm (with cap)
Used for processing brain, liver, kidney, lung, spleen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.