Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Improved geometry
- Short distance of 0.3 mm between external well bottom and microplate skirt bottom
- Facilitates changing objectives during microscopy
Specifications
Specifications
| Format | Standard |
| Description | Model: Sensoplate Plus |
| Array | 32 x 48 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |