Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isovaleryl chloride

Catalog No. 15742225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-742-225G 25 g
15-742-2500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-742-225G Supplier Sigma Aldrich Supplier No. 15742225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isovaleryl chloride was used in the synthesis of:
  • Furanodictines a and b
  • Tetrapeptide amide, s-benzyl-L-cysteinyl-L-prolyl-L-leucylglycinamide
  • (+)-blastmycinone
  • (r)- and (s)-2-methyl-4-octanol, aggregation pheromone of some sugarcane weevils

Specifications

CAS 108-12-3
Density 0.989 g/mL (at 25°C (literature))
Boiling Point 115°C to 117°C (lit.)
Molecular Formula C5H9ClO
Refractive Index n20/D 1.416 (literature)
Linear Formula (CH3)2 CHCH2COCl
MDL Number MFCD00000738
Quantity 25 g
Synonym 3-Methylbutyryl chloride
Molecular Weight (g/mol) 120.58
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.