Learn More
Mopec™ #15C Sterile Scalpel Blades
SDP
Catalog No. 22445392 Shop All Mopec Products
Change view
Click to view available options
Blade Material:
Carbon Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Material
22-445-392 Carbon Steel
22-445-677 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-445-392 Supplier Mopec™ Supplier No. AJ175
Add to Cart
Add to Cart
High-quality, sterile blades used for anatomic pathology or autopsy.
  • Individually wrapped
  • Fits #3, #3L and #5 scalpel handles
Specifications
Blade Material Carbon Steel
Product Type Scalpel
Size # 15
Disposable Single-use
Sterility Sterile
Graduated No