Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 16-Doxyl-Stearic Acid, Free Radical

Catalog No. 25359625MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-359-625MG 25 mg
25-359-610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-359-625MG Supplier Sigma Aldrich Supplier No. 25359625MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

16-Doxyl-Stearic Acid, Free Radical

Specifications

CAS 53034-38-1
Melting Point 47°C to 55°C
Molecular Formula C22H42NO4
Linear Formula C22H42NO4
MDL Number MFCD00010066
Quantity 25 mg
Synonym 16-SASL; 16DOXYL; 16NS; 2-(14-Carboxytetradecyl)-2-ethyl-4,4-dimethyl-3-oxazolidinyloxy, free radical
Molecular Weight (g/mol) 384.57 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.