Learn More
PIP 16 in. Black Polyblend Steel Toe and Shank Boot
Catalog No. 17986481 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
13
14
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-986-481 6
17-986-482 7
17-986-483 8
17-986-484 9
17-986-485 10
17-986-486 11
17-986-487 12
17-986-488 13
17-986-489 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-986-481 Supplier PIP Supplier No. 383890/6
Add to Cart
Add to Cart
Steel Toe Polyblend Waterproof Boot.Steel Shank. Pre-Formed Insole. ASTM F2413. Made in USA.
  • Less toe spring allows for more surface contact for improved channeling of water for better traction, slip resistance and comfort
  • Steel Shank provides improved support and ladder stability
  • Energy absorbing tread adds all-day comfort
  • Tough outsole for good abrasion resistance. Flexible upper material stays supple in cold temperatures to make walking easier
  • Midsole is designed to utilize additional material in the standard area of the shank providing increased support and comfort
  • Lower profile 'kick off' ledge on back of boots for ease of removal
  • Meets ASTM F3445-21 for Slip Resistance
  • Meets ASTM F2413-18 M/I/C Impact and Compression
  • Made in the USA
  • Prop 65 Compliant
  • Resistant to certain acids and alkalies, fats, oils hydrocarbons and other chemicals
Specifications
Color Black
Material PVC
Size 6
Toe Material Steel
Gender Unisex
Waterproof Yes
Product Type Safety Boot