Learn More
PIP 16 in. Black Polyblend Steel Toe and Shank Boot
Catalog No. 17986489 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
13
14
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-986-489 14
17-986-481 6
17-986-482 7
17-986-483 8
17-986-484 9
17-986-485 10
17-986-486 11
17-986-487 12
17-986-488 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-986-489 Supplier PIP Supplier No. 383890/14
  • $535.00
    $255.65 / Case of 6
    Save $279.35
    52% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Steel Toe Polyblend Waterproof Boot.Steel Shank. Pre-Formed Insole. ASTM F2413. Made in USA.
  • Less toe spring allows for more surface contact for improved channeling of water for better traction, slip resistance and comfort
  • Steel Shank provides improved support and ladder stability
  • Energy absorbing tread adds all-day comfort
  • Tough outsole for good abrasion resistance. Flexible upper material stays supple in cold temperatures to make walking easier
  • Midsole is designed to utilize additional material in the standard area of the shank providing increased support and comfort
  • Lower profile 'kick off' ledge on back of boots for ease of removal
  • Meets ASTM F3445-21 for Slip Resistance
  • Meets ASTM F2413-18 M/I/C Impact and Compression
  • Made in the USA
  • Prop 65 Compliant
  • Resistant to certain acids and alkalies, fats, oils hydrocarbons and other chemicals
Specifications
Color Black
Material PVC
Size 14
Toe Material Steel
Gender Unisex
Waterproof Yes
Product Type Safety Boot