Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Succinonitrile

Catalog No. 16096225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-096-225G 25 g
16-096-2100G 100 g
16-096-2500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-096-225G Supplier Sigma Aldrich Supplier No. 16096225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Succinonitrile

Specifications

CAS 110-61-2
Melting Point 50°C to 54°C (lit.)
Density 0.985 g/mL (at 25°C (literature))
Boiling Point 265°C to 267°C (lit.)
Molecular Formula C4H4N2
Linear Formula NCCH2CH2CN
Quantity 25 g
MDL Number MFCD00001949
Synonym 1,2-Dicyanoethane; Butanedinitrile
Molecular Weight (g/mol) 80.09
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.