Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromo-1-propanol

Catalog No. 16716925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-716-925G 25 g
16-716-95G 5 g
16-716-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-716-925G Supplier Sigma Aldrich Supplier No. 16716925G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reaction between 3-bromo-1-propanol, phenol and a series of phenols having substituents in 4-position has been studied in micellar media and in microemulsions based on cationic or a nonionic surfactant.Recently used as a grafting agent in the synthesis of recyclable reagents for Swern oxidation

Specifications

CAS 627-18-9
Density 1.537 g/mL (at 25°C (literature))
Boiling Point 62°C (5 mmHg)
Molecular Formula C3H7BrO
Refractive Index n20/D 1.488 (literature)
Linear Formula Br(CH2)3 OH
MDL Number MFCD00002942
Quantity 25 g
Synonym Trimethylene bromohydrin
Molecular Weight (g/mol) 138.99
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.