Learn More
Thomas Scientific 16" Deep Upright Freezer Drawer Racks for 6 x 2" Boxes and 30 x 50ml Tubes, 16-1/2"D x 9-11/16"H x 5-1/2"W, 2 Drawers.
SDP
Supplier:  Thomas Scientific 1148X26
Encompass
Rack w/2 Drawers for 4 Standard 2" Cardboard Boxes&I Drawer for Storage Bottles 8 Boxes Plus 80 Tubes (15ml) 8 Boxes Plus 39 Tubes (50ml) Outside Dimensions:22 x 9 11/16 x 5 1/2 inch in ( 559 x 246 x 140
Catalog No. 50-196-027
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.