Learn More
Market Lab 16mm Tube Racks for 10mL Tubes, Medium, Holds 72 tubes, 5"W x 9.5"L x 2.5"H
Supplier:  Market Lab 7216
Encompass_Preferred
Holds 72 tubes, strong and durable, chemical-resistant epoxy coating, and autoclavable for easy cleaning.
Catalog No. NC1781340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.