Learn More
LAKE CHARLES MFG 16x150mm Test Tubes, Polystyrene, Round BTM, 20ml Vol, Neon Yellow, Karter Scientific 400C1 (Case of 1000)
Supplier:  LAKE CHARLES MFG 400C1
Encompass_Preferred
Cylindrical Test Tube, manufactured from high quality polystyrene. PS Tubes are not autoclavable or dishwasher safe but are chemically resistant and clear like glass. Polystyrene has the properties of glass but at a much lower price point. Since polystyrene is not as hard as glass it will have minor scratches. Colors is Neon Yellow.
Catalog No. NC2180889
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.