Learn More
17-AAG (HSP90 inhibitor), Boston BioChem™
Specifications
Specifications
| Assay Percent Range | >98% |
| Quantity | 100 μg |
| Formula Weight | 585.7 |
| Assay | by HPLC |
| Chemical Name or Material | 17-AAG (HSP90 inhibitor), Synthetic |
Safety and Handling
| Recommended Storage | Store the unopened product at -20 to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.