Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17-DMAG hydrochloride, Tocris Bioscience™

Catalog No. 26101
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-101 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-101 Supplier R&D Systems Supplier No. 2610/1
This item is not returnable. View return policy

Water-soluble Hsp90 inhibitor

Water-soluble analog of 17-AAG (Cat. No. 1515) and geldanamycin (Cat. No. 1368). Binds the ATP binding site of Hsp90 and inhibits its chaperone activity. Displays more potent antitumor activity than 17-AAG (mean GI50 values are 53 and 123 nM for 17-DMAG and 17-AAG respectively).

Chemical Identifiers

CAS 467214-21-7
Molecular Formula C32H49ClN4O8
Molecular Weight (g/mol) 653.214
InChI Key DFSYBWLNYPEFJK-ZOKRJHFSSA-N
Synonym 17-Demethoxy-17-[[2-(dimethylamino)ethyl]amino]geldanamycin hydrochloride
PubChem CID 131954774
IUPAC Name [(3R,5S,6R,7S,8E,10S,11S)-21-[2-(dimethylamino)ethylamino]-6-hydroxy-5,11-dimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate;hydrochloride
SMILES CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)NCCN(C)C)C)OC)OC(=O)N)C)C)O)OC.Cl

Specifications

Quantity 1 mg
Formula Weight 653.21
Percent Purity >99%
Chemical Name or Material 17-DMAG hydrochloride
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.