Learn More
Medchemexpress LLC 17-hydroxyisolathyrol | 93551-00-9 | MFCD32178668 | 99.3% | 350.45 g·mol-1 | C20H30O5 | 10 MG

Supplier: Medchemexpress LLC HYN413210MG
17-Hydroxyisolathyrol is a macrocyclic lathyrol derivative isolated from seeds of Euphorbia lathyris, supplied for research use and characterization in biochemical and pharmacological studies.
- High purity: 99.3% (HPLC)
- Well-characterized: CAS 93551-00-9
- Chemical formula C20H30O5 and molecular weight 350.45 g·mol-1
- Available in small research quantities suitable for screening and assays
- Soluble in common organic solvents such as DMSO for in vitro work
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.