Learn More
- Home
- Products
- Animal Care and Research
- Animal Surgical Devices
- Animal Surgical Equipment
- Stoelting™ 1700 Series Gas-Tight Microliter Syringe
Stoelting™ 1700 Series Gas-Tight Microliter Syringe
Catalog No. 10000899
Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Bevel tip
Blunt tip
Luer tip
Size:
10 μL syringe
10 μL syringe; 26 ga
100 μL syringe
100 μL syringe; 22 ga
25 μL syringe
25 μL syringe; 22 ga
250 μL syringe
250 μL syringe; 22 ga
50 μL syringe
50 μL syringe; 22 ga
500 μL syringe
500 μL syringe; 22 ga
Includes:
—
Removable Needle
21 product options available for selection
| Catalog No. | Model | Size | Includes |
|---|---|---|---|
| 10-000-899 | Bevel tip | 100 μL syringe; 22 ga | — |
| 10-000-896 | Bevel tip | 10 μL syringe; 26 ga | — |
| 10-000-901 | Blunt tip | 10 μL syringe; 26 ga | — |
| 10-000-889 | Luer tip | 10 μL syringe | — |
| 10-000-909 | Bevel tip | 100 μL syringe; 22 ga | Removable Needle |
| 10-000-904 | Blunt tip | 100 μL syringe; 22 ga | — |
| 10-000-915 | Blunt tip | 100 μL syringe; 22 ga | Removable Needle |
| 10-000-892 | Luer tip | 100 μL syringe | — |
| 10-000-897 | Bevel tip | 25 μL syringe; 22 ga | — |
| 10-000-902 | Blunt tip | 25 μL syringe; 22 ga | — |
| 10-000-890 | Luer tip | 25 μL syringe | — |
| 10-000-900 | Bevel tip | 250 μL syringe; 22 ga | — |
| 10-000-910 | Bevel tip | 250 μL syringe; 22 ga | Removable Needle |
| 10-000-905 | Blunt tip | 250 μL syringe; 22 ga | — |
| 10-000-916 | Blunt tip | 250 μL syringe; 22 ga | Removable Needle |
| 10-000-893 | Luer tip | 250 μL syringe | — |
| 10-000-903 | Blunt tip | 50 μL syringe; 22 ga | — |
| 10-000-898 | Bevel tip | 50 μL syringe; 22 ga | — |
| 10-000-891 | Luer tip | 50 μL syringe | — |
| 10-000-917 | Blunt tip | 500 μL syringe; 22 ga | Removable Needle |
| 10-000-894 | Luer tip | 500 μL syringe | — |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
Description
Ideal for dispensing volumes from 1 μL up to 500 μL.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Size | 100 μL syringe; 22 ga |
| Model | Bevel tip |
| Includes | — |